Product References
Cissus quadrangularis (Hadjod) Inhibits RANKL-Induced Osteoclastogenesis and Augments Bone Health in an Estrogen-Deficient Preclinical Model of Osteoporosis Via Modulating the Host Osteoimmune System.
Cells
Azam Z,Sapra L,Baghel K,Sinha N,Gupta RK,Soni V,Saini C,Mishra PK,Srivastava RK
Published figure using CD5 monoclonal antibody (Product # 14-0051-82) in Flow Cytometry
Wed Jan 04 00:00:00 EST 2023
IL-33 induces thymic involution-associated naive T cell aging and impairs host control of severe infection.
Nature communications
Xu L,Wei C,Chen Y,Wu Y,Shou X,Chen W,Lu D,Sun H,Li W,Yu B,Wang X,Zhang X,Yu Y,Lei Z,Tang R,Zhu J,Li Y,Lu L,Zhou H,Zhou S,Su C,Chen X
Published figure using CD5 monoclonal antibody (Product # 14-0051-82) in Flow Cytometry
Sat Nov 12 00:00:00 EST 2022
The Influence of B Cell Depletion Therapy on Naturally Acquired Immunity to Streptococcus pneumoniae.
Frontiers in immunology
Ercoli G,Ramos-Sevillano E,Nakajima R,de Assis RR,Jasinskas A,Goldblatt D,Felgner P,Weckbecker G,Brown J
Published figure using CD5 monoclonal antibody (Product # 14-0051-82) in Flow Cytometry
Tue Jun 22 00:00:00 EDT 2021
Antigen-presenting ILC3 regulate T cell-dependent IgA responses to colonic mucosal bacteria.
The Journal of experimental medicine
Melo-Gonzalez F,Kammoun H,Evren E,Dutton EE,Papadopoulou M,Bradford BM,Tanes C,Fardus-Reid F,Swann JR,Bittinger K,Mabbott NA,Vallance BA,Willinger T,Withers DR,Hepworth MR
14-0051-82 was used in Flow Cytometry to study the mechanisms that modulate the magnitude and quality of T cell-dependent IgA responses.
Mon Apr 01 00:00:00 EDT 2019
Decreased immunoglobulin G in brain regions of elder female APOE4-TR mice accompany with Aβ accumulation.
Immunity & ageing : I & A
Zhang L,Xu J,Gao J,Chen P,Yin M,Zhao W
14-0051-82 was used in Flow Cytometry to determine the alterations of IgG, CD19, and Aβ in various brain regions of uninfected male and female APOE3- and APOE4-TR mice at the age of 3 and 10 months to elucidate impacts of AD risk factors on alterations of brain IgG.
Thu Mar 31 00:00:00 EDT 2022
Inhibition of T cell activation and function by the adaptor protein CIN85.
Science signaling
Kong MS,Hashimoto-Tane A,Kawashima Y,Sakuma M,Yokosuka T,Kometani K,Onishi R,Carpino N,Ohara O,Kurosaki T,Phua KK,Saito T
14-0051-82 was used in Flow Cytometry to show that CIN85 (Cbl-interacting protein of 85 kDa) limited T cell responses to T cell receptor stimulation.
Tue Feb 05 00:00:00 EST 2019
Host Tumor Suppressor p18INK4c Functions as a Potent Cell-Intrinsic Inhibitor of Murine Gammaherpesvirus 68 Reactivation and Pathogenesis.
Journal of virology
Niemeyer BF,Oko LM,Medina EM,Oldenburg DG,White DW,Cool CD,Clambey ET,van Dyk LF
Published figure using CD5 monoclonal antibody (Product # 14-0051-82) in Flow Cytometry
Thu Mar 15 00:00:00 EDT 2018
Bruton's Tyrosine Kinase Is Not Essential for B Cell Survival beyond Early Developmental Stages.
Journal of immunology (Baltimore, Md. : 1950)
Nyhoff LE,Clark ES,Barron BL,Bonami RH,Khan WN,Kendall PL
14-0051 was used in Flow cytometry/Cell sorting to raise the possibility that Btk may also be expendable for survival of mature human B cells, therefore requiring prolonged dosing to be effective, and that success of BTK inhibitors may depend in part on off-target effects.
Sun Apr 01 00:00:00 EDT 2018
Identification of Distinct Unmutated Chronic Lymphocytic Leukemia Subsets in Mice Based on Their T Cell Dependency.
Frontiers in immunology
Pal Singh S,de Bruijn MJW,de Almeida MP,Meijers RWJ,Nitschke L,Langerak AW,Pillai SY,Stadhouders R,Hendriks RW
14-0051 was used in Flow cytometry/Cell sorting to provide evidence for two distinct unmutated CLL subsets with a specific transcriptional signature: one is T cell-independent and B-1 cell-derived while the other arises upon antigen stimulation in the context of T-cell help.
Mon Sep 30 00:00:00 EDT 2019
Differential requirement of kindlin-3 for T cell progenitor homing to the non-vascularized and vascularized thymus.
eLife
Moretti FA,Klapproth S,Ruppert R,Margraf A,Weber J,Pick R,Scheiermann C,Sperandio M,Fässler R,Moser M
Published figure using CD5 monoclonal antibody (Product # 14-0051-82) in Flow Cytometry
Thu Sep 06 00:00:00 EDT 2018
CTCF-Mediated Enhancer-Promoter Interaction Is a Critical Regulator of Cell-to-Cell Variation of Gene Expression.
Molecular cell
Ren G,Jin W,Cui K,Rodrigez J,Hu G,Zhang Z,Larson DR,Zhao K
Published figure using CD5 monoclonal antibody (Product # 14-0051-82) in Flow Cytometry
Thu Sep 21 00:00:00 EDT 2017
CD73 regulates anti-inflammatory signaling between apoptotic cells and endotoxin-conditioned tissue macrophages.
Cell death and differentiation
Murphy PS,Wang J,Bhagwat SP,Munger JC,Janssen WJ,Wright TW,Elliott MR
14-0051 was used in Flow cytometry/Cell sorting to show that, in RAW264.7 cells, ectopic expression of both A2a and CD73 was required for TNF suppression by apoptotic cells.
Wed Mar 01 00:00:00 EST 2017
Naïve helper T cells with high CD5 expression have increased calcium signaling.
PloS one
Freitas CMT,Hamblin GJ,Raymond CM,Weber KS
Published figure using CD5 monoclonal antibody (Product # 14-0051-82) in Flow Cytometry
Tue Sep 26 00:00:00 EDT 2017
Human and Mouse Hematopoietic Stem Cells Are a Depot for Dormant Mycobacterium tuberculosis.
PloS one
Tornack J,Reece ST,Bauer WM,Vogelzang A,Bandermann S,Zedler U,Stingl G,Kaufmann SH,Melchers F
14-0051-82 was used in Flow Cytometry to conclude that LT-pHSCs, together with MSCs of Mtb-infected humans and mice serve as a hitherto unappreciated quiescent cellular depot for Mtb during latent TB infection.
Tue Sep 05 00:00:00 EDT 2017
Arginase 1 is an innate lymphoid-cell-intrinsic metabolic checkpoint controlling type 2 inflammation.
Nature immunology
Monticelli LA,Buck MD,Flamar AL,Saenz SA,Tait Wojno ED,Yudanin NA,Osborne LC,Hepworth MR,Tran SV,Rodewald HR,Shah H,Cross JR,Diamond JM,Cantu E,Christie JD,Pearce EL,Artis D
14-0051 was used in Flow cytometry/Cell sorting to identify Arg1 as a key regulator of group 2 innate lymphoid cell bioenergetics that control proliferative capacity and pro-inflammatory functions.
Wed Jun 01 00:00:00 EDT 2016
Chronic interleukin-1 exposure drives haematopoietic stem cells towards precocious myeloid differentiation at the expense of self-renewal.
Nature cell biology
Pietras EM,Mirantes-Barbeito C,Fong S,Loeffler D,Kovtonyuk LV,Zhang S,Lakshminarasimhan R,Chin CP,Techner JM,Will B,Nerlov C,Steidl U,Manz MG,Schroeder T,Passegué E
14-0051 was used in Flow cytometry/Cell sorting to identify a critical regulatory circuit that tailors HSC responses to acute needs, and is likely to underlie deregulated blood homeostasis in chronic inflammation conditions.
Wed Jun 01 00:00:00 EDT 2016
LNK/SH2B3 regulates IL-7 receptor signaling in normal and malignant B-progenitors.
The Journal of clinical investigation
Cheng Y,Chikwava K,Wu C,Zhang H,Bhagat A,Pei D,Choi JK,Tong W
14-0051 was used in Flow cytometry/Cell sorting to investigate how LNK regulates normal B cell development or promotes leukemogenesis.
Fri Apr 01 00:00:00 EDT 2016
Agonist-Mediated Activation of STING Induces Apoptosis in Malignant B Cells.
Cancer research
Tang CH,Zundell JA,Ranatunga S,Lin C,Nefedova Y,Del Valle JR,Hu CC
14-0051 was used in Flow cytometry/Cell sorting to investigate the role of STING in inducing apoptosis in malignant B cells.
Fri Apr 15 00:00:00 EDT 2016
The BAFF receptor TACI controls IL-10 production by regulatory B cells and CLL B cells.
Leukemia
Saulep-Easton D,Vincent FB,Quah PS,Wei A,Ting SB,Croce CM,Tam C,Mackay F
14-0051 was used in Flow cytometry/Cell sorting to uncover a major targetable pathway important for the generation of regulatory B cells that is detrimental to immunity in chronic lymphocytic leukemia.
Fri Jan 01 00:00:00 EST 2016
Protective Role for B-1b B Cells and IgM in Obesity-Associated Inflammation, Glucose Intolerance, and Insulin Resistance.
Arteriosclerosis, thrombosis, and vascular biology
Harmon DB,Srikakulapu P,Kaplan JL,Oldham SN,McSkimming C,Garmey JC,Perry HM,Kirby JL,Prohaska TA,Gonen A,Hallowell P,Schirmer B,Tsimikas S,Taylor AM,Witztum JL,McNamara CA
14-0051 was used in Flow cytometry/Cell sorting to investigate the role of B-cell functions in obesity-induced metabolic dysfunction.
Fri Apr 01 00:00:00 EDT 2016
Group 2 innate lymphoid cells utilize the IRF4-IL-9 module to coordinate epithelial cell maintenance of lung homeostasis.
Mucosal immunology
Mohapatra A,Van Dyken SJ,Schneider C,Nussbaum JC,Liang HE,Locksley RM
14-0051 was used in Flow cytometry/Cell sorting to study how ILC2s link alveolar function to regulation of airway flow, revealing a key interaction between resident lymphoid and structural cells that might underlie similar organisational hierarchies in other organs.
Fri Jan 01 00:00:00 EST 2016
Blood Stem Cell Activity Is Arrested by Th1-Mediated Injury Preventing Engraftment following Nonmyeloablative Conditioning.
Journal of immunology (Baltimore, Md. : 1950)
Müller AM,Florek M,Kohrt HE,Küpper NJ,Filatenkov A,Linderman JA,Hadeiba H,Negrin RS,Shizuru JA
14-0051 was used in Flow cytometry/Cell sorting to shed new light on the effects of CD4+ T cells on HSC biology.
Tue Nov 15 00:00:00 EST 2016
NFATc1 supports imiquimod-induced skin inflammation by suppressing IL-10 synthesis in B cells.
Nature communications
Alrefai H,Muhammad K,Rudolf R,Pham DA,Klein-Hessling S,Patra AK,Avots A,Bukur V,Sahin U,Tenzer S,Goebeler M,Kerstan A,Serfling E
Published figure using CD5 monoclonal antibody (Product # 14-0051-82) in Flow Cytometry
Wed May 25 00:00:00 EDT 2016
CXXC finger protein 1 is critical for T-cell intrathymic development through regulating H3K4 trimethylation.
Nature communications
Cao W,Guo J,Wen X,Miao L,Lin F,Xu G,Ma R,Yin S,Hui Z,Chen T,Guo S,Chen W,Huang Y,Liu Y,Wang J,Wei L,Wang L
14-0051 was used in Flow cytometry/Cell sorting to investigate whether the CpG-binding protein Cxxc1 plays a role in thymocyte development.
Mon May 23 00:00:00 EDT 2016
The atypical chemokine receptor ACKR2 suppresses Th17 responses to protein autoantigens.
Immunology and cell biology
Hansell CA,MacLellan LM,Oldham RS,Doonan J,Chapple KJ,Anderson EJ,Linington C,McInnes IB,Nibbs RJ,Goodyear CS
14-0051 was used in Flow cytometry/Cell sorting to investigate the role of ACKR2 in autoimmunity, showing that it suppresses Th17 responses to protein autoantigens.
Sun Feb 01 00:00:00 EST 2015
B cell Rab7 mediates induction of activation-induced cytidine deaminase expression and class-switching in T-dependent and T-independent antibody responses.
Journal of immunology (Baltimore, Md. : 1950)
Pone EJ,Lam T,Lou Z,Wang R,Chen Y,Liu D,Edinger AL,Xu Z,Casali P
Published figure using CD5 monoclonal antibody (Product # 14-0051-82) in Flow Cytometry
Wed Apr 01 00:00:00 EDT 2015
CCR7-dependent trafficking of RORγ⁺ ILCs creates a unique microenvironment within mucosal draining lymph nodes.
Nature communications
Mackley EC,Houston S,Marriott CL,Halford EE,Lucas B,Cerovic V,Filbey KJ,Maizels RM,Hepworth MR,Sonnenberg GF,Milling S,Withers DR
14-0051 was used in Flow cytometry/Cell sorting to investigate whether group 3 innate lymphoid cells migrate from their mucosal and peripheral sites to local draining secondary lymphoid tissues.
Fri Jan 09 00:00:00 EST 2015
Overexpression of RhoH Permits to Bypass the Pre-TCR Checkpoint.
PloS one
Tamehiro N,Oda H,Shirai M,Suzuki H
14-0051 was used in Flow cytometry/Cell sorting to indicate that an excess amount of RhoH is able to initiate pre-TCR signalling in the absence of pre-TCR complexes.
Mon Apr 25 00:00:00 EDT 2016
In utero depletion of fetal hematopoietic stem cells improves engraftment after neonatal transplantation in mice.
Blood
Derderian SC,Togarrati PP,King C,Moradi PW,Reynaud D,Czechowicz A,Weissman IL,MacKenzie TC
14-0051 was used in Flow cytometry/Cell sorting to develop a successful foetal conditioning strategy associated with minimal toxicity.
Thu Aug 07 00:00:00 EDT 2014
Inhibition of ER stress-associated IRE-1/XBP-1 pathway reduces leukemic cell survival.
The Journal of clinical investigation
Tang CH,Ranatunga S,Kriss CL,Cubitt CL,Tao J,Pinilla-Ibarz JA,Del Valle JR,Hu CC
14-0051 was used in Flow cytometry/Cell sorting to investigate the role of ER stress response in chronic lymphocytic leukaemia, showing that inhibition of ER stress-associated IRE-1/XBP-1 pathway reduces leukemic cell survival.
Sun Jun 01 00:00:00 EDT 2014
In vivo functional mapping of the conserved protein domains within murine Themis1.
Immunology and cell biology
Zvezdova E,Lee J,El-Khoury D,Barr V,Akpan I,Samelson L,Love PE
14-0051 was used in Flow cytometry/Cell sorting to investigate the functional importance of different Themis1 domains.
Mon Sep 01 00:00:00 EDT 2014
B cell-specific loss of Lyn kinase leads to autoimmunity.
Journal of immunology (Baltimore, Md. : 1950)
Lamagna C,Hu Y,DeFranco AL,Lowell CA
14-0051-82 was used in Flow Cytometry to demonstrate that B cell-intrinsic Lyn-dependent signalling pathways regulate B cell homeostasis and activation.
Sat Feb 01 00:00:00 EST 2014
Recognition of tumor cells by Dectin-1 orchestrates innate immune cells for anti-tumor responses.
eLife
Chiba S,Ikushima H,Ueki H,Yanai H,Kimura Y,Hangai S,Nishio J,Negishi H,Tamura T,Saijo S,Iwakura Y,Taniguchi T
14-0051 was used in Flow cytometry/Cell sorting to show that recognition of tumor cells by Dectin-1 receptors leads to activation of the tumoricidal activity of NK cells.
Fri Aug 22 00:00:00 EDT 2014
A canonical to non-canonical Wnt signalling switch in haematopoietic stem-cell ageing.
Nature
Florian MC,Nattamai KJ,Dörr K,Marka G,Uberle B,Vas V,Eckl C,Andrä I,Schiemann M,Oostendorp RA,Scharffetter-Kochanek K,Kestler HA,Zheng Y,Geiger H
14-0051 was used in Immunocytochemistry to evaluate the role of Wnt signalling in the ageing of haematopoietic stem cells in mice.
Thu Nov 21 00:00:00 EST 2013
A canonical to non-canonical Wnt signalling switch in haematopoietic stem-cell ageing.
Nature
Florian MC,Nattamai KJ,Dörr K,Marka G,Uberle B,Vas V,Eckl C,Andrä I,Schiemann M,Oostendorp RA,Scharffetter-Kochanek K,Kestler HA,Zheng Y,Geiger H
14-0051 was used in Immunocytochemistry to evaluate the role of Wnt signalling in the ageing of haematopoietic stem cells in mice.
Thu Nov 21 00:00:00 EST 2013
T cell-positive selection uses self-ligand binding strength to optimize repertoire recognition of foreign antigens.
Immunity
Mandl JN,Monteiro JP,Vrisekoop N,Germain RN
14-0051 was used in Flow cytometry/Cell sorting to provide evidence for a connection between thymic recognition events and foreign antigen-driven T cell responses.
Thu Feb 21 00:00:00 EST 2013
Natural killer cells are required for extramedullary hematopoiesis following murine cytomegalovirus infection.
Cell host & microbe
Jordan S,Ruzsics Z,Mitrović M,Baranek T,Arapović J,Krmpotić A,Vivier E,Dalod M,Jonjić S,Dölken L,Koszinowski UH
14-0051 was used in Flow cytometry/Cell sorting to investigate the underlying mechanisms of extramedullary hematopoiesis.
Wed May 15 00:00:00 EDT 2013
Cutaneous immunosurveillance and regulation of inflammation by group 2 innate lymphoid cells.
Nature immunology
Roediger B,Kyle R,Yip KH,Sumaria N,Guy TV,Kim BS,Mitchell AJ,Tay SS,Jain R,Forbes-Blom E,Chen X,Tong PL,Bolton HA,Artis D,Paul WE,Fazekas de St Groth B,Grimbaldeston MA,Le Gros G,Weninger W
14-0051 was used in Flow cytometry/Cell sorting to report the identification in normal mouse dermis of an abundant, phenotypically unique group 2 innate lymphoid cell subset that depended on interleukin 7 and constitutively produced IL-13.
Sat Jun 01 00:00:00 EDT 2013
Histone demethylase Lsd1 represses hematopoietic stem and progenitor cell signatures during blood cell maturation.
eLife
Kerenyi MA,Shao Z,Hsu YJ,Guo G,Luc S,O'Brien K,Fujiwara Y,Peng C,Nguyen M,Orkin SH
14-0051 was used in Flow cytometry/Cell sorting to study how Lsd1-mediated concurrent repression of enhancer and promoter activity of stem and progenitor cell genes is a pivotal epigenetic mechanism required for proper hematopoietic maturation.
Tue Jun 18 00:00:00 EDT 2013
Unique features of naive CD8+ T cell activation by IL-2.
Journal of immunology (Baltimore, Md. : 1950)
Cho JH,Kim HO,Kim KS,Yang DH,Surh CD,Sprent J
14-0051 was used in in vitro experiments to demonstrate that IL-2 elicits a unique signaling pattern associated with strong lymphocyte-specific protein tyrosine kinase/JAK3-dependent activation of the PI3K/AKT pathway.
Sun Dec 01 00:00:00 EST 2013
Critical role of SAP in progression and reactivation but not maintenance of T cell-dependent humoral immunity.
Molecular and cellular biology
Zhong MC,Veillette A
14-0051 was used in Flow cytometry/Cell sorting to explore the therapeutic potential of blocking the SLAM-associated protein (SAP) for human autoimmune diseases.
Fri Mar 01 00:00:00 EST 2013
Molecular definition of the identity and activation of natural killer cells.
Nature immunology
Bezman NA,Kim CC,Sun JC,Min-Oo G,Hendricks DW,Kamimura Y,Best JA,Goldrath AW,Lanier LL
14-0051 was used in Flow cytometry/Cell sorting to systematically define the transcriptome of mouse natural killer cells in several contexts.
Mon Oct 01 00:00:00 EDT 2012
Unexpected role for MHC II-peptide complexes in shaping CD8 T-cell expansion and differentiation in vivo.
Proceedings of the National Academy of Sciences of the United States of America
Do JS,Valujskikh A,Vignali DA,Fairchild RL,Min B
14-0051 was used in Flow cytometry/Cell sorting to reveal a previously undefined MHC II-dependent regulation that tunes CD8 T-cell reactivity.
Tue Jul 31 00:00:00 EDT 2012
PIDDosome-independent tumor suppression by Caspase-2.
Cell death and differentiation
Manzl C,Peintner L,Krumschnabel G,Bock F,Labi V,Drach M,Newbold A,Johnstone R,Villunger A
14-0051 was used in Flow cytometry/Cell sorting to investigate the role of PIDD, RIP-1 kinase and Nemo/IKKγ in oncogenesis induced by DNA damage or oncogenic stress in gene-ablated mice.
Mon Oct 01 00:00:00 EDT 2012
Rescue of tumor-infiltrating lymphocytes from activation-induced cell death enhances the antitumor CTL response in CD5-deficient mice.
Journal of immunology (Baltimore, Md. : 1950)
Tabbekh M,Franciszkiewicz K,Haouas H,Lécluse Y,Benihoud K,Raman C,Mami-Chouaib F
14-0051-81 was used in Flow Cytometry to compare the T cell response to the B16F10 melanoma engrafted into CD5-deficient and wild-type C57BL/6 mice.
Fri Jul 01 00:00:00 EDT 2011
Anti-CD180 (RP105) activates B cells to rapidly produce polyclonal Ig via a T cell and MyD88-independent pathway.
Journal of immunology (Baltimore, Md. : 1950)
Chaplin JW,Kasahara S,Clark EA,Ledbetter JA
14-0051 was used in Flow cytometry/Cell sorting to study the effect of injection of anti-CD180 mAb into mice on the production of immunoglobulins.
Sat Oct 15 00:00:00 EDT 2011
L2pB1: a new player in autoimmunity.
Molecular immunology
Zhong X,Rothstein TL
14-0051 was used in Flow cytometry/Cell sorting to review the features of PD-L2 positive B1 cells and their contribution to autoimmunity.
Wed Jun 01 00:00:00 EDT 2011
Suppression of B-cell lymphomagenesis by the BH3-only proteins Bmf and Bad.
Blood
Frenzel A,Labi V,Chmelewskij W,Ploner C,Geley S,Fiegl H,Tzankov A,Villunger A
14-0051 was used in Flow cytometry/Cell sorting to demonstrate the potential of BH3-only proapoptotic Bcl-2 family members to regulate excessive lymphocyte proliferation.
Thu Feb 04 00:00:00 EST 2010
B-cell homeostasis requires complementary CD22 and BLyS/BR3 survival signals.
International immunology
Smith SH,Haas KM,Poe JC,Yanaba K,Ward CD,Migone TS,Tedder TF
14-0051 was used in Flow cytometry/Cell sorting to support a model whereby B cells must integrate varied signals, including those from the BLyS- BR3 axis, CD22-ligand binding and antigen-receptor-mediated tonic signaling for their homeostatic maintenance in vivo.
Sun Aug 01 00:00:00 EDT 2010
Oncogenic Kras initiates leukemia in hematopoietic stem cells.
PLoS biology
Sabnis AJ,Cheung LS,Dail M,Kang HC,Santaguida M,Hermiston ML,Passegué E,Shannon K,Braun BS
14-0051 was used in Flow cytometry/Cell sorting to conclude that MPD-initiating activity is restricted to the HSC compartment in Kras(G12D) mice.
Tue Mar 17 00:00:00 EDT 2009
Developmental acquisition of the Lyn-CD22-SHP-1 inhibitory pathway promotes B cell tolerance.
Journal of immunology (Baltimore, Md. : 1950)
Gross AJ,Lyandres JR,Panigrahi AK,Prak ET,DeFranco AL
14-0051 was used in Flow cytometry/Cell sorting to indicate that the Lyn-CD22 inhibitory pathway largely becomes operational as B cells mature, and is critical for the maintenance of tolerance in the B cell compartment.
Fri May 01 00:00:00 EDT 2009
Murine isolated lymphoid follicles contain follicular B lymphocytes with a mucosal phenotype.
American journal of physiology. Gastrointestinal and liver physiology
Wang C,McDonald KG,McDonough JS,Newberry RD
14-0051 was used in Flow cytometry/Cell sorting to indicate that murine ILFs contain a polyclonal population of follicular B-2 B lymphocytes with a phenotype similar to PP B lymphocytes.
Sun Oct 01 00:00:00 EDT 2006